Thermo Scientific Chemicals

Lead(II) nitrate, 99%, Thermo Scientific Chemicals

Catalog number: A16345.0E
2500 g, Each
Thermo Scientific Chemicals

Lead(II) nitrate, 99%, Thermo Scientific Chemicals

Catalog number: A16345.0E
2500 g, Each
Quantity
Catalog number: A16345.0E
also known as A16345-0E
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10099-74-8
IUPAC Name
λ²-lead(2+) dinitrate
Molecular Formula
N2O6Pb
InChI Key
RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Complexometric Titration
≥98.5 to ≤101.5%
Appearance (Color)
White

Description

Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.

Solubility
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.

Notes
Incompatible with strong reducing agents, organic materials and powdered metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text