Lead(II) nitrate, 99%, Thermo Scientific Chemicals
Lead(II) nitrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) nitrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16345.36
also known as A16345-36
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.40 
Save 9.75 (14%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.40 
Save 9.75 (14%)
Add to cart
Lead(II) nitrate, 99%, Thermo Scientific Chemicals
Catalog numberA16345.36
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.40 
Save 9.75 (14%)
-
Add to cart
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Complexometric Titration≥98.5 to ≤101.5%
Appearance (Color)White
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.

Solubility
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.

Notes
Incompatible with strong reducing agents, organic materials and powdered metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Used to promote the formation of N-9-phenylfluorenyl protected derivatives from amino acids or their esters and 9-bromo-9-phenylfluorene: J. Org. Chem., 54, 3824 (1989); Org. Synth. Coll., 9, 344 (1998).
  2. Deschenes, G.; Lastra, R.; Brown, J. R.; Jina, S.; May, O.; Ghali, E. Effect of lead nitrate on cyanidation of gold ores: progress on the study of the mechanisms. Miner. Eng. 2000, 13 (12), 1263-1279.
  3. Sayiner, B. Influence of Lead Nitrate on Cyanide Leaching of Gold and Silver from Turkish Gold Ores. Physicochem. Probl. Miner. Process. 2014, 50 (2), 507-514.