Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A16346.0B
1000 g, Each
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A16346.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16346.0B
also known as A16346-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-13-1
IUPAC Name
λ²-cobalt(2+) hexahydrate dichloride
Molecular Formula
Cl2CoH12O6
InChI Key
GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Appearance (Color)
Red-purple or purple
Assay (Titration ex Chloride)
≥97.5 to ≤102.5% (as hexahydrate)
Water Content (Karl Fischer Titration)
42.2-48.4% (5.25-6.75 waters)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

Solubility
Soluble in water, ether, acetone and alcohol.

Notes
Incompatible with strong oxidizing agents and metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text