Thermo Scientific Chemicals

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A16347.14
25 g, Each
Thermo Scientific Chemicals

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A16347.14
25 g, Each
Quantity
Catalog number: A16347.14
also known as A16347-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1530-45-6
IUPAC Name
(2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular Formula
C22H22BrO2P
InChI Key
VJVZPTPOYCJFNI-UHFFFAOYSA-M
SMILES
[Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Form
Powder or crystalline powder
Assay (Titration ex Bromide)
≥97.5% to ≤102.5%
Water Content (Karl Fischer Titration)
<1.0%

Description

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text