Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16502.30
250 g, Each
Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16502.30
250 g, Each
Quantity
Catalog number: A16502.30
also known as A16502-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-34-3
IUPAC Name
3,5-dinitrobenzoic acid
Molecular Formula
C7H4N2O6
InChI Key
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Appearance (Color)
White to pale cream to pale yellow to pale brown
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text