Thermo Scientific Chemicals

Fast Green FCF

Catalog number: A16520.06
5 g, Each
Thermo Scientific Chemicals

Fast Green FCF

Catalog number: A16520.06
5 g, Each
Quantity
Catalog number: A16520.06
also known as A16520-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2353-45-9
IUPAC Name
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular Formula
C37H34N2Na2O10S3
InChI Key
XJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Appearance (Color)
Dark red to dark brown
Dye Strength
>95.0 to <105.0% (dry weight basis)
Solution Test
Clear orange in cHCl : Clear blue in 10% NaOH
Form
Powder
Loss on Drying
=< 15% (200?C, constant weight)

Description

Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson's trichrome. Further, it is utilized in tissue stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson′s trichrome. Further, it is utilized in tissue stain.

Solubility
Soluble in water, glycerin, glycol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text