Fast Green FCF
Thermo Scientific Chemicals

Fast Green FCF

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A16520.22
Price (USD)/ Each
258.65
Online exclusive
287.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
258.65
Online exclusive
287.00 
Save 28.35 (10%)
Add to cart
Fast Green FCF
Catalog numberA16520.22
Price (USD)/ Each
258.65
Online exclusive
287.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2353-45-9
IUPAC Namedisodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular FormulaC37H34N2Na2O10S3
InChI KeyXJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to dark brown
Dye Strength>95.0 to <105.0% (dry weight basis)
Solution TestClear orange in cHCl : Clear blue in 10% NaOH
FormPowder
Loss on Drying=< 15% (200?C, constant weight)
View more
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson's trichrome. Further, it is utilized in tissue stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson′s trichrome. Further, it is utilized in tissue stain.

Solubility
Soluble in water, glycerin, glycol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Dalavi, D. K.; Kamble, A.; Bhopate, D. P.; Mahajan, P. G.; Kolekar, G. B.; Patil, S. R. TNPs as a novel fluorescent sensor for the selective recognition of fast green FCF: a spectrofluorimetric approach. RSC Adv. 2015, 5 (85), 69371-69377.
  2. Li, Q.; Tan, X.; Zheng, X.; Tang, W.; Yang, J. A novel method for the determination of fast green in grape wine based on resonance Rayleigh scattering. J. Mol. Struct. 2015, 1100, 14-20.