Thermo Scientific Chemicals

Dibenzofuran, 98%, Thermo Scientific Chemicals

Catalog number: A16521.22
100 g, Each
Thermo Scientific Chemicals

Dibenzofuran, 98%, Thermo Scientific Chemicals

Catalog number: A16521.22
100 g, Each
Quantity
Catalog number: A16521.22
also known as A16521-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-64-9
IUPAC Name
8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Formula
C12H8O
InChI Key
TXCDCPKCNAJMEE-UHFFFAOYSA-N
SMILES
O1C2=C(C=CC=C2)C2=C1C=CC=C2
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Melting Point (clear melt)
79.0-85.0°C

Description

Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text