Thermo Scientific Chemicals

Methyl carbamate, 99%, Thermo Scientific Chemicals

Catalog number: A16523.22
100 g, Each
Thermo Scientific Chemicals

Methyl carbamate, 99%, Thermo Scientific Chemicals

Catalog number: A16523.22
100 g, Each
Quantity
Catalog number: A16523.22
also known as A16523-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
598-55-0
Appearance (Color)
White
Assay (GC)
≥98.5%
Melting Point (clear melt)
51-57°C
Form
Crystals or crystalline powder
Identification (FTIR)
Conforms

Description

Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

Solubility
Soluble in water(20g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text