Thermo Scientific Chemicals

D-(-)-Salicin, 99%, Thermo Scientific Chemicals

Catalog number: A16524.14
25 g, Each
Thermo Scientific Chemicals

D-(-)-Salicin, 99%, Thermo Scientific Chemicals

Catalog number: A16524.14
25 g, Each
Quantity
Catalog number: A16524.14
also known as A16524-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138-52-3
IUPAC Name
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
Molecular Formula
C13H18O7
InChI Key
NGFMICBWJRZIBI-UJPOAAIJSA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC=CC=C2CO)[C@H](O)[C@@H](O)[C@@H]1O
Assay (HPLC)
≥98.5%
Optical Rotation
-59.5? to -63.5? (c=5 in Water)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream

Description

Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alkalies, pyridine, glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.

Notes
Store in cool place. Light and heat sensitive. Air senstive and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text