Thermo Scientific Chemicals

2,3-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A16526.14
25 g, Each
Thermo Scientific Chemicals

2,3-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A16526.14
25 g, Each
Quantity
Catalog number: A16526.14
also known as A16526-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56737-75-8
IUPAC Name
(2,3-dimethylphenyl)hydrazine hydrochloride
Molecular Formula
C8H13ClN2
InChI Key
UKAYNIJBIXNDDJ-UHFFFAOYSA-N
SMILES
Cl.CC1=C(C)C(NN)=CC=C1
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder or flakes
Appearance (Color)
Pale cream to cream to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text