BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.
Solubility
Soluble in DMF (0.1 g/mL). Reacts with water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Horng-Long Tyan,; Kung-Hwa Wei,; Tsung-Eong Hsieh. Mechanical properties of clay-polyimide (BTDA-ODA) nanocomposites via ODA-modified organoclay. Journal of Polymer Science Part B: Polymer Physics. 2008, 38(44),2873-2878.
James F. Dezern. Synthesis and characterization of BTDA-based polyamide-imides. Journal of Polymer Science Part A: Polymer Chemistry. 1988, 26 (9), 2157-2169.