Thermo Scientific Chemicals

7-Hydroxy-4-(trifluoromethyl)coumarin, 98%, Thermo Scientific Chemicals

Catalog number: A16537.06
5 g, Each
Thermo Scientific Chemicals

7-Hydroxy-4-(trifluoromethyl)coumarin, 98%, Thermo Scientific Chemicals

Catalog number: A16537.06
5 g, Each
Quantity
Catalog number: A16537.06
also known as A16537-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
575-03-1
IUPAC Name
7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
Molecular Formula
C10H5F3O3
InChI Key
CCKWMCUOHJAVOL-UHFFFAOYSA-N
SMILES
OC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream or pale pink to pink
Melting Point (clear melt)
182.0-188.0?C
Assay (Silylated GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text