Thermo Scientific Chemicals

Cupferron, 97+%, Thermo Scientific Chemicals

Catalog number: A16551.14
25 g, Each
Thermo Scientific Chemicals

Cupferron, 97+%, Thermo Scientific Chemicals

Catalog number: A16551.14
25 g, Each
Quantity
Catalog number: A16551.14
also known as A16551-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135-20-6
IUPAC Name
N-nitroso-N-phenylhydroxylamine amine
Molecular Formula
C6H9N3O2
InChI Key
NJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILES
N.ON(N=O)C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream or pale yellow
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%
Form
Crystals or powder or crystalline powder

Description

Reagent for determination of Ce, Cu, Fe, Sn, and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text