Thermo Scientific Chemicals

2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16587.18
50 g, Each
Thermo Scientific Chemicals

2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16587.18
50 g, Each
Quantity
Catalog number: A16587.18
also known as A16587-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3142-72-1
IUPAC Name
(2E)-2-methylpent-2-enoate
Molecular Formula
C6H9O2
InChI Key
JJYWRQLLQAKNAD-SNAWJCMRSA-M
SMILES
CC\C=C(/C)C([O-])=O
Assay (GC)
≥98.5%
Appearance (Color)
White/colorless
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Refractive Index
1.4580-1.4610 @ 20?C (melt)
Form
Low melting solid or clear colorless liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text