2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals
2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A16587.30
also known as A16587-30
Price (USD)/ Each
492.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
492.00
Add to cart
2-Methyl-2-pentenoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA16587.30
Price (USD)/ Each
492.00
-
Add to cart
Chemical Identifiers
CAS3142-72-1
IUPAC Name(2E)-2-methylpent-2-enoate
Molecular FormulaC6H9O2
InChI KeyJJYWRQLLQAKNAD-SNAWJCMRSA-M
SMILESCC\C=C(/C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White/colorless
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Refractive Index1.4580-1.4610 @ 20?C (melt)
FormLow melting solid or clear colorless liquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only