Thermo Scientific Chemicals

2-Fluoro-4-methylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A16591.14
25 g, Each
Thermo Scientific Chemicals

2-Fluoro-4-methylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A16591.14
25 g, Each
Quantity
Catalog number: A16591.14
also known as A16591-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
452-80-2
IUPAC Name
2-fluoro-4-methylaniline
Molecular Formula
C7H8FN
InChI Key
ZQEXBVHABAJPHJ-UHFFFAOYSA-N
SMILES
CC1=CC=C(N)C(F)=C1
Form
Liquid
Refractive Index
1.5310-1.5340 @ 20?C
Appearance (Color)
Clear colorless or yellow to orange or pale red to pale brown
Assay (GC)
≥98.5%

Description

2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

Solubility
Not miscible in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text