1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A16597.14
also known as A16597-14
Price (USD)/ Each
283.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
283.00
Add to cart
1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals
Catalog numberA16597.14
Price (USD)/ Each
283.00
-
Add to cart
Chemical Identifiers
CAS33863-76-2
IUPAC Name1-bromo-3-chloro-5-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyGGMDFPMASIXEIR-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5470-1.5510 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only