Thermo Scientific Chemicals

2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals

Catalog number: A16606.06
5 g, Each
Thermo Scientific Chemicals

2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals

Catalog number: A16606.06
5 g, Each
Quantity
Catalog number: A16606.06
also known as A16606-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
524-95-8
IUPAC Name
2-aminoethyl diphenylborinate
Molecular Formula
C14H16BNO
InChI Key
BLZVCIGGICSWIG-UHFFFAOYSA-N
SMILES
NCCOB(C1=CC=CC=C1)C1=CC=CC=C1
Melting Point (clear melt)
188.0-195.0?C
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

Solubility
Soluble in ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text