Thermo Scientific Chemicals

D-Pantothenic acid calcium salt, 98%, Thermo Scientific Chemicals

Catalog number: A16609.36
500 g, Each
Thermo Scientific Chemicals

D-Pantothenic acid calcium salt, 98%, Thermo Scientific Chemicals

Catalog number: A16609.36
500 g, Each
Quantity
Catalog number: A16609.36
also known as A16609-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-08-6
IUPAC Name
calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
Molecular Formula
C18H32CaN2O10
InChI Key
FAPWYRCQGJNNSJ-DXHDTSSINA-L
SMILES
[Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
Form
Powder
Heavy Metals
≤20ppm
Comment
Material sourced in the U.S. and in other countries
Impurity content
≤1.0% related substances (non-U.S. sourced material)
Optical Rotation
+25.0 to +27.5° (c=5, water)

Description

Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

Solubility
Soluble in water.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text