4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals
4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A16610.14
also known as A16610-14
Price (USD)/ Each
184.65
Online exclusive
205.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
184.65
Online exclusive
205.00 
Save 20.35 (10%)
Add to cart
4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberA16610.14
Price (USD)/ Each
184.65
Online exclusive
205.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS69954-66-1
IUPAC Namehydrogen 4-(dimethylamino)butanoic acid chloride
Molecular FormulaC6H14ClNO2
InChI KeyRDTALXUBMCLWBB-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)148.0-154.0?C
FormPowder
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.
RUO – Research Use Only
Xue-Bo Yin; Bin Qi; Xuping Sun; Xiurong Yang and Erkang Wang. 4-(Dimethylamino)butyric Acid Labeling for Electrochemiluminescence Detection of Biological Substances by Increasing Sensitivity with Gold Nanoparticle Amplification. Anal. Chem.2005, 77 (11), 3525-3530.