Thermo Scientific Chemicals

Boric acid, 98%, Thermo Scientific Chemicals

Catalog number: A16624.0B
1000 g, Each
Thermo Scientific Chemicals

Boric acid, 98%, Thermo Scientific Chemicals

Catalog number: A16624.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16624.0B
also known as A16624-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-35-3
IUPAC Name
boric acid
Molecular Formula
BH3O3
InChI Key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES
OB(O)O
Comment
Material sourced in the U.S. and in other countries
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

Solubility
Soluble in glycerol, ether, alcohol, methanol, liquid ammonia, acetone (slightly), water (6.35 g/100 cc) at 30°C, water (27.6 g/100 cc) at 100°C, water (50 g/L) at 21°C, and water (1 g / 18 ml cold water).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text