3-Benzoylpropionic acid, 98+%, Thermo Scientific Chemicals
3-Benzoylpropionic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Benzoylpropionic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16636.14
also known as A16636-14
Price (USD)/ Each
56.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
56.30
Add to cart
3-Benzoylpropionic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA16636.14
Price (USD)/ Each
56.30
-
Add to cart
Chemical Identifiers
CAS2051-95-8
IUPAC Name4-oxo-4-phenylbutanoic acid
Molecular FormulaC10H10O3
InChI KeyKMQLIDDEQAJAGJ-UHFFFAOYSA-N
SMILESOC(=O)CCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)113.0-119.0?C
FormPowder
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only