Ethyl propionate, 99%, Thermo Scientific Chemicals
Ethyl propionate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl propionate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number A16650.AU
also known as A16650-AU
Price (USD)/ Each
62.10
-
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (USD)/ Each
62.10
Add to cart
Ethyl propionate, 99%, Thermo Scientific Chemicals
Catalog numberA16650.AU
Price (USD)/ Each
62.10
-
Add to cart
Chemical Identifiers
CAS105-37-3
IUPAC Nameethyl propanoate
Molecular FormulaC5H10O2
InChI KeyFKRCODPIKNYEAC-UHFFFAOYSA-N
SMILESCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3830-1.3865 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only