Thermo Scientific Chemicals

Diphenylphosphinamide, 98%, Thermo Scientific Chemicals

Catalog number: A16657.06
5 g, Each
Thermo Scientific Chemicals

Diphenylphosphinamide, 98%, Thermo Scientific Chemicals

Catalog number: A16657.06
5 g, Each
Quantity
Catalog number: A16657.06
also known as A16657-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5994-87-6
IUPAC Name
[amino(phenyl)phosphoroso]benzene
Molecular Formula
C12H12NOP
InChI Key
RIGIWEGXTTUCIQ-UHFFFAOYSA-N
SMILES
NP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
159.0-165.0?C

Description

Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

Solubility
Soluble in chloroform, nitromethane, nitrobenzene.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text