Thermo Scientific Chemicals

Oleic acid, tech. 90%, Thermo Scientific Chemicals

Catalog number: A16663.0J
5000 mL, Each
Thermo Scientific Chemicals

Oleic acid, tech. 90%, Thermo Scientific Chemicals

Catalog number: A16663.0J
5000 mL, Each
Quantity
Catalog number: A16663.0J
also known as A16663-0J
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-80-1
IUPAC Name
(9E)-octadec-9-enoic acid
Molecular Formula
C18H34O2
InChI Key
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILES
CCCCCCCC\C=C\CCCCCCCC(O)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (Silylated GC)
≥88.0% (non-U.S. specification)
Assay from Suppliers CofA
≥88.0% (U.S. specification)
Acid Value
≥196 (U.S specification)

Description

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

Solubility
Miscible with ethanol, ether, acetone, chloroform, dimethyl formamide and dimethyl sulfoxide.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and aluminum.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text