Thermo Scientific Chemicals

β-Alanine, 98%

Catalog number: A16665.0B
1000 g, Each
Thermo Scientific Chemicals

β-Alanine, 98%

Catalog number: A16665.0B
1000 g, Each
Quantity
Catalog number: A16665.0B
also known as A16665-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-95-9
IUPAC Name
3-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
UCMIRNVEIXFBKS-UHFFFAOYSA-N
SMILES
NCCC(O)=O
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.

Solubility
Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone.

Notes
Slightly hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text