Thermo Scientific Chemicals

2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals

Catalog number: A16667.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals

Catalog number: A16667.14
25 g, Each
Quantity
Catalog number: A16667.14
also known as A16667-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-46-1
IUPAC Name
(carbamothioylamino)thiourea
Molecular Formula
C2H6N4S2
InChI Key
KCOYHFNCTWXETP-UHFFFAOYSA-N
SMILES
NC(=S)NNC(N)=S
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Assay (HPLC)
≥96.0%

Description

2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

Solubility
Soluble in hot water, pyridine, triethylamine and dimethylformamide. Insoluble in cold water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text