Thermo Scientific Chemicals

2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%, Thermo Scientific Chemicals

Catalog number: A16679.03
1 g, Each
Thermo Scientific Chemicals

2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%, Thermo Scientific Chemicals

Catalog number: A16679.03
1 g, Each
Quantity
Catalog number: A16679.03
also known as A16679-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
846-63-9
IUPAC Name
2-(naphthalen-1-yl)-5-phenyl-1,3-oxazole
Molecular Formula
C19H13NO
InChI Key
WWVFJJKBBZXWFV-UHFFFAOYSA-N
SMILES
O1C(=CN=C1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Appearance (Color)
Yellow to green
Assay (GC)
>99.0%
Form
Crystalline powder
Specific Absorbance (E 1%/1cm)
>21500 (1%, 1cm)
lambda-max (UV-Vis)
331-335nm (in 1,4-Dioxane)

Description

Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

Solubility
Sparingly Soluble (4.1E-4 g/L) (25°C). Soluble most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text