Thermo Scientific Chemicals

2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%, Thermo Scientific Chemicals

Catalog number: A16680.18
50 g, Each
Thermo Scientific Chemicals

2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%, Thermo Scientific Chemicals

Catalog number: A16680.18
50 g, Each
Quantity
Catalog number: A16680.18
also known as A16680-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
815-60-1
IUPAC Name
(2R,4S)-2,4-dibromopentan-3-one
Molecular Formula
C5H8Br2O
InChI Key
UOPIOAUZQKSZRO-ZXZARUISSA-N
SMILES
C[C@@H](Br)C(=O)[C@H](C)Br
Form
Liquid
Refractive Index
1.5025-1.5075 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

Solubility
Slightly soluble in water(50 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text