Thermo Scientific Chemicals

Quinhydrone, 97%, Thermo Scientific Chemicals

Catalog number: A16681.22
100 g, Each
Thermo Scientific Chemicals

Quinhydrone, 97%, Thermo Scientific Chemicals

Catalog number: A16681.22
100 g, Each
Quantity
Catalog number: A16681.22
also known as A16681-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-34-3
IUPAC Name
benzene-1,4-diol; cyclohexa-2,5-diene-1,4-dione
Molecular Formula
C12H10O4
InChI Key
BDJXVNRFAQSMAA-UHFFFAOYSA-N
SMILES
OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
Melting Point (clear melt)
165-175?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Dark green
Assay (HPLC)
≥96.0%

Description

Used to measure the hydrogen ion concentration (pH) of a solution in a chemical experiment. It provides an alternative to the commonly used glass electrode in a pH meter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to measure the hydrogen ion concentration (pH) of a solution in a chemical experiment. It provides an alternative to the commonly used glass electrode in a pH meter.

Solubility
Slightly soluble in water 4 g/L (20°C).

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text