3-Noradamantanecarboxylic acid, 98%, Thermo Scientific Chemicals
3-Noradamantanecarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Noradamantanecarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number A16683.01
also known as A16683-01
Price (USD)/ Each
37.40
-
Add to cart
Quantity:
0.25 g
Request bulk or custom format
Price (USD)/ Each
37.40
Add to cart
3-Noradamantanecarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog numberA16683.01
Price (USD)/ Each
37.40
-
Add to cart
Chemical Identifiers
CAS16200-53-6
IUPAC Name(1R,5S)-tricyclo[3.3.1.0³,⁷]nonane-3-carboxylate
Molecular FormulaC10H13O2
InChI KeyRXUUYFUQAGICCD-NLZKFAMNSA-M
SMILES[O-]C(=O)C12C[C@@H]3CC1C[C@H](C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)105.0-111.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only