Thermo Scientific Chemicals

Tetra-n-butylammonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: A16688.06
5 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: A16688.06
5 g, Each
Quantity
Catalog number: A16688.06
also known as A16688-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
429-42-5
IUPAC Name
tetrabutylazanium; tetrafluoroboranuide
Molecular Formula
C16H36BF4N
InChI Key
NNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White to pale cream
Purity (DSC)
≥98.5%
Melting Point (clear melt)
157-163?C
Form
Crystals or crystalline powder or powder

Description

As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

Solubility
Slightly soluble in Water, methanol( 0.1 g/mL), ethanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text