Thermo Scientific Chemicals

3-Benzoylpyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A16695.14
25 g, Each
Thermo Scientific Chemicals

3-Benzoylpyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A16695.14
25 g, Each
Quantity
Catalog number: A16695.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5424-19-1
IUPAC Name
3-benzoylpyridine
Molecular Formula
C12H9NO
InChI Key
RYMBAPVTUHZCNF-UHFFFAOYSA-N
SMILES
O=C(C1=CC=CC=C1)C1=CC=CN=C1
Form
Fused solid or clear liquid as melt
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream to yellow
Melting Point (clear melt)
34.0-41.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text