Thermo Scientific Chemicals

3-Bromo-4-methoxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A16698.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-4-methoxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A16698.06
5 g, Each
Quantity
Catalog number: A16698.06
also known as A16698-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117572-79-9
Assay (GC)
≥98.5%
Melting Point (clear melt)
119.0-125.0?C
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder

Description

3-Bromo-4-methoxybenzonitrile is used to prepare 3-bromo-2-iodo-4-methoxy-benzonitrile. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxybenzonitrile is used to prepare 3-bromo-2-iodo-4-methoxy-benzonitrile. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text