Thermo Scientific Chemicals

Alizarin Complexone, may cont. up to 10% water, Thermo Scientific Chemicals

Catalog number: A16699.03
1 g, Each
Thermo Scientific Chemicals

Alizarin Complexone, may cont. up to 10% water, Thermo Scientific Chemicals

Catalog number: A16699.03
1 g, Each
Quantity
Catalog number: A16699.03
also known as A16699-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3952-78-1
IUPAC Name
2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid
Molecular Formula
C19H15NO8
InChI Key
PWIGYBONXWGOQE-UHFFFAOYSA-N
SMILES
OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=CC=CC=C1C2=O
Appearance (Color)
Orange to brown
Form
powder
Water Content (Karl Fischer Titration)
≤ 10.0%
lambda-max (UV-Vis)
425 - 430nm (in methanol)

Description

Alizarin is the main ingredient for the manufacture of the madder lake pigments known to painters as Rose madder and Alizarin crimson. A notable use of alizarin in modern times is as a staining agent in biological research because it stains free calcium and certain calcium compounds a red or light purple color. Alizarin Red is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin is the main ingredient for the manufacture of the madder lake pigments known to painters as Rose madder and Alizarin crimson. A notable use of alizarin in modern times is as a staining agent in biological research because it stains free calcium and certain calcium compounds a red or light purple color. Alizarin Red is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text