9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Shi, S.; Allonas, X.; Croutxé-Barghorn, C.; Chemtob, A. Activation of the sol-gel process by visible light-emitting diodes (LEDs) for the synthesis of inorganic films. New J. Chem. 2015, 39 (7), 5686-5693.
Volatron, F.; Noël, J. M.; Rinfray, C.; Decorse, P.; Combellas, C.; Kanoufi, F.; Proust, A. Electron transfer properties of a monolayer of hybrid polyoxometalates on silicon. J. Mater. Chem. C 2015, 3 (24), 6266-6275.