4-Fluorocinnamic acid, 98+%, Thermo Scientific Chemicals
4-Fluorocinnamic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorocinnamic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16707.06
also known as A16707-06
Price (USD)/ Each
40.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
40.60
Add to cart
4-Fluorocinnamic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA16707.06
Price (USD)/ Each
40.60
-
Add to cart
Chemical Identifiers
CAS459-32-5
IUPAC Name(2E)-3-(4-fluorophenyl)prop-2-enoic acid
Molecular FormulaC9H7FO2
InChI KeyISMMYAZSUSYVQG-ZZXKWVIFSA-N
SMILESOC(=O)\C=C\C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)206.0-213.0?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only