Phenyltrichlorosilane, 97%, Thermo Scientific Chemicals
Phenyltrichlorosilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyltrichlorosilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A16713.36
also known as A16713-36
Price (USD)/ Each
59.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
59.00
Add to cart
Phenyltrichlorosilane, 97%, Thermo Scientific Chemicals
Catalog numberA16713.36
Price (USD)/ Each
59.00
-
Add to cart
Chemical Identifiers
CAS98-13-5
IUPAC Nametrichloro(phenyl)silane
Molecular FormulaC6H5Cl3Si
InChI KeyORVMIVQULIKXCP-UHFFFAOYSA-N
SMILESCl[Si](Cl)(Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.5215-1.5265 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only