N-Benzyloxycarbonyl-L-alanine, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-alanine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-alanine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A16716.22
also known as A16716-22
Price (USD)/ Each
225.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
225.00
Add to cart
N-Benzyloxycarbonyl-L-alanine, 98%, Thermo Scientific Chemicals
Catalog numberA16716.22
Price (USD)/ Each
225.00
-
Add to cart
Chemical Identifiers
CAS1142-20-7
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular FormulaC11H13NO4
InChI KeyTYRGLVWXHJRKMT-QMMMGPOBSA-N
SMILESC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder
Optical Rotation-14.5 ? 1? (c=2 in acetic acid)
Assay (Aqueous acid-base Titration)>97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only