Thermo Scientific Chemicals

1-Pentene, 97%, Thermo Scientific Chemicals

Catalog number: A16723.22
100 g, Each
Thermo Scientific Chemicals

1-Pentene, 97%, Thermo Scientific Chemicals

Catalog number: A16723.22
100 g, Each
Quantity
Catalog number: A16723.22
also known as A16723-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-67-1
IUPAC Name
pent-1-ene
Molecular Formula
C5H10
InChI Key
YWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILES
CCCC=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
≥96.0%

Description

1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents, halogens and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text