Thermo Scientific Chemicals

1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals

Catalog number: A16724.01
0.25 g, Each
Thermo Scientific Chemicals

1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals

Catalog number: A16724.01
0.25 g, Each
Quantity
Catalog number: A16724.01
also known as A16724-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
498-07-7
Assay from Supplier's CofA
≥98.5%
Form
Powder or crystals or crystalline powder
Appearance (Color)
White

Description

1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

Solubility
Soluble in Ethanol and Water.

Notes
Store at -20°C. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text