Thermo Scientific Chemicals

3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A16729.14
25 g, Each
Thermo Scientific Chemicals

3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A16729.14
25 g, Each
Quantity
Catalog number: A16729.14
also known as A16729-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-33-2
IUPAC Name
3,5-dinitrobenzoyl chloride
Molecular Formula
C7H3ClN2O5
InChI Key
NNOHXABAQAGKRZ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(=CC(=C1)C(Cl)=O)[N+]([O-])=O
Form
Crystals or needles or powder or crystalline powder
Appearance (Color)
White to cream to yellow to brown
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Melting Point
67-72?C, may be slight turbidity
Identification (FTIR)
Conforms

Description

Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text