2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals
2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16733.06
also known as A16733-06
Price (USD)/ Each
66.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
66.60
Add to cart
2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA16733.06
Price (USD)/ Each
66.60
-
Add to cart
Chemical Identifiers
CAS2040-04-2
IUPAC Name1-(2,6-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O3
InChI KeyXEUGKOFTNAYMMX-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)66-71?C
FormCrystals or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only