Thermo Scientific Chemicals

4-Methylcyclohexanol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A16734.AK
250 mL, Each
Thermo Scientific Chemicals

4-Methylcyclohexanol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A16734.AK
250 mL, Each
Quantity
Catalog number: A16734.AK
also known as A16734-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
589-91-3
IUPAC Name
4-methylcyclohexan-1-ol
Molecular Formula
C7H14O
InChI Key
MQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILES
CC1CCC(O)CC1
Refractive Index
1.4565-1.4605 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text