Lead(II) acetate, basic, Thermo Scientific Chemicals
Lead(II) acetate, basic, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) acetate, basic, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16763.0E
also known as A16763-0E
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
Add to cart
Lead(II) acetate, basic, Thermo Scientific Chemicals
Catalog numberA16763.0E
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1335-32-6
IUPAC Nametris(λ²-lead(2+)) diacetate tetrahydroxide
Molecular FormulaC4H10O8Pb3
InChI KeyVLOJXAQYHIVPFI-UHFFFAOYSA-H
SMILES[OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)>33.0% basic lead as PbO
Assay (unspecified)Pb content 74.5% - 77.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only