Cesium sulfate, 99%, Thermo Scientific Chemicals
Cesium sulfate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium sulfate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16767.14
also known as A16767-14
Price (USD)/ Each
55.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
55.00
Add to cart
Cesium sulfate, 99%, Thermo Scientific Chemicals
Catalog numberA16767.14
Price (USD)/ Each
55.00
-
Add to cart
Chemical Identifiers
CAS10294-54-9
IUPAC Namedicaesium(1+) sulfate
Molecular FormulaCs2O4S
InChI KeyFLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.5%
Appearance (Color)White
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

Solubility
Soluble in water. (1820 g/L) at 20°C

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
RUO – Research Use Only

General References:

  1. D W Kufe; P P Major; E M Egan; E Loh. 5-Fluoro-2'-deoxyuridine incorporation in L1210 DNA. Journal of Biological Chemistry.1981, 56 (1), 83-89.
  2. P J Roughley; R J White. The use of cesium sulphate density gradient centrifugation to analyse proteoglycans from human articular cartilages of different ages. Biochimica et Biophysica Acta.1983, 759 (1-2), 58-66.