CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).
Solubility
Miscible with water, glycolether and ketones.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only
General References:
- S. Richard Turner. Development of amorphous copolyesters based on 1, 4-cyclohexanedimethanol. ournal of Polymer Science Part A: Polymer Chemistry. 2004, 42(24),5847-5852.
- John Scheirs,; Timothy E. Long,; S. R. Turner, R. W. Seymour,; J. R. Dombroski. Amorphous and Crystalline Polyesters Based on 1,4-Cyclohexanedimethanol. Modern Polyesters: Chemistry and Technology of Polyesters and Copolyesters.