Evans Blue
Thermo Scientific Chemicals

Evans Blue

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A16774.09
also known as A16774-09
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
Add to cart
Evans Blue
Catalog numberA16774.09
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Chemical Identifiers
CAS314-13-6
IUPAC Nametetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC34H24N6Na4O14S4
InChI KeyKBNIFDASRCWYGC-GXNXWABVSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and/or Crystals
Specific Absorbance (E 1%/1cm)≥600 (Lambda max in distilled water, pH 7.0)
Appearance (Color)Dark brown to dark red to purple
Assay (Dye)>80%
Color ChangeDark blue to purple on addition of 0.1N NaOH
View more
Evans Blue is used for estimation of blood volume and in double-labeling procedure for studying axonal branching. It acts as an inhibitor of L-glutamate and kainate receptor-mediated currents. It is also a P2X-selective purinoceptor antagonist. It is also useful in molecular biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yang, J.; Jiang, Y.; Guo, Y. Measuring Blood-brain-barrier Permeability Using Evans Blue in Mice. Bio-protocol. 2015, 5 (15), 1548.
  2. Herz, J.; Johnson, K. R.; McGavern, D. B. Therapeutic antiviral T cells noncytopathically clear persistently infected microglia after conversion into antigen-presenting cells. J Exp. Med. 2015, 212 (8), 1153-1169.