3-Nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
3-Nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Nitrobenzyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A16775.18
also known as A16775-18
Price (USD)/ Each
134.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
134.00
Add to cart
3-Nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA16775.18
Price (USD)/ Each
134.00
-
Add to cart
Chemical Identifiers
CAS619-23-8
IUPAC Name1-(chloromethyl)-3-nitrobenzene
Molecular FormulaC7H6ClNO2
InChI KeyAPGGSERFJKEWFG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(CCl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)42.0-48.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only